Polyethylene glycol monomethyl ether, M.W. 350

Supplier: Thermo Scientific Chemicals

41560-22 41560-36
CAAA41560-22EA 110.6 CAD
CAAA41560-22 CAAA41560-36
Polyethylene glycol monomethyl ether, M.W. 350
Polyethylene glycol monomethyl ether

CAS No.: 9004-74-4 Molecular Formula: CH3(OCH2CH2)nOH Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.09 Appearance: Colorless to pale yellow MDL No.: MFCD00084416

Formula: CH₃(OCH₂CH₂)nOH
Density: 1,09
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR