3-(3,4-Dihydroxyphenyl)-DL-alanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DL-β-3,4-Dihydroxyphenyl-Alanine, DL-DOPA, 3,4-Dihydroxy-DL-phenylalanine

41535-06 41535-14
CAAA41535-06EA 128.93 CAD
CAAA41535-06 CAAA41535-14 CAAA41535-22
3-(3,4-Dihydroxyphenyl)-DL-alanine 98%
3-(3,4-Dihydroxyphenyl)-DL-alanine

Purity: 98% CAS No.: 63-84-3 Molecular Formula: C9H11NO4 Formula Weight: 197.19 Physical Form: Crystalline Odor: Odorless Merck Reference: 14,3420 MDL No.: MFCD00063060

Formula: C₉H₁₁NO₄
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063060
CAS Number: 63-84-3
EINECS: 200-566-0
Merck Index: 14,3420

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR