Di-n-octyl phthalate 98%
Supplier: Thermo Scientific Chemicals
41522-AC
41522-AE
41522-AP
CAAA41522-ACEA
95.43
CAD
CAAA41522-AC
CAAA41522-AE
CAAA41522-AP
Di-n-octyl phthalate 98%
Di-n-octyl phthalate
Applications: Plasticizer
Purity: 98%
CAS No.: 117-84-0
Molecular Formula: C24H38O4
Formula Weight: 390.56
Flash Point: 219°(426°F)
Physical Form: Liquid
Density: 0.98
Appearance: Colorless to plae yellow
Melting Point: -25°
Boiling Point: 380°
Merck Reference: 14,2864
MDL No.: MFCD00015292
Plasticizer
Formula:
C₂₄H₃₈O₄ Boiling Pt: 380 °C Melting Pt: –25 °C Density: 0.98 Flash Pt: 219 °C (426 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015292 CAS Number: 117-84-0 EINECS: 204-214-7 Merck Index: 14,2864 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...