Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution ≥98% in vinyl terminated polydimethylsiloxane

Supplier: Thermo Scientific Chemicals

Synonyms: Karstedt Catalyst, Platinum(0) - 1,3-Divinyltetramethyldisiloxane Complex

41508-03 41508-06 41508-14
CAAA41508-03EA 91.32 CAD
CAAA41508-03 CAAA41508-06 CAAA41508-14
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution ≥98% in vinyl terminated polydimethylsiloxane
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex

Applications: Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst


CAS No.: 68478-92-2 Molecular Formula: (C8H18OSi2)1.5Pt Formula Weight: 474.68 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.984 Sensitivity: Moisture Sensitive Appearance: Orange MDL No.: MFCD00151662


Applications: Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst

Formula: C₈H₁₈OPtSi₂
Density: 0.984
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151662
CAS Number: 68478-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR