Yttrium aluminum oxide ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
41491-14
41491-22
CAAA41491-14EA
312.84
CAD
CAAA41491-14
CAAA41491-22
Yttrium aluminum oxide ≥99.9% (REO, rare earth oxide basis)
Yttrium aluminum oxide
Applications: Rare earth element doped yttrium aluminum oxide is used in solid state lasers.
Purity: 99.9%
CAS No.: 12005-21-9
Molecular Formula: Y3Al5O12
Formula Weight: 593.62
Physical Form: Powder
Density: 4.65
Refractive Index: 1.833
Melting Point: 1760°
MDL No.: MFCD00213958
Applications: Rare earth element doped yttrium aluminum oxide is used in solid state lasers.
Formula:
Y₃Al₅O₁₂ Melting Pt: 1760 °C Density: 4,65 Storage Temperature: Ambient |
MDL Number:
MFCD00213958 CAS Number: 12005-21-9 EINECS: 234-465-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...