Molybdenum(V) isopropoxide ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41484-02
41484-04
CAAA41484-02EA
131.14
CAD
CAAA41484-02
CAAA41484-04
Molybdenum(V) isopropoxide ≥99%
Molybdenum(V) isopropoxide
Applications: Precursor used in sol-gel process
Purity: 99+%
CAS No.: 209733-38-0
Molecular Formula: C15H35MoO5
Formula Weight: 391.04
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown to black
UN No.: 3181
MDL No.: MFCD00210636
Applications: Precursor used in sol-gel process
Formula:
C₁₅H₃₅MoO₅ MW: 391.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210636 CAS Number: 209733-38-0 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...