Ethyl glycolate ≥95%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate
41475-09
41475-18
CAAA41475-09EA
88.95
CAD
CAAA41475-09
CAAA41475-18
Ethyl glycolate ≥95%
Ethyl glycolate
Purity: 95%
CAS No.: 623-50-7
Molecular Formula: C4H8O3
Formula Weight: 104.11
Flash Point: 61°(143°F)
Physical Form: Liquid
Density: 1.10
Refractive Index: 1.4190
Appearance: Colorless
Boiling Point: 158-159°
MDL No.: MFCD00021970
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 158…159 °C Density: 1,10 Flash Pt: 61 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021970 CAS Number: 623-50-7 EINECS: 210-798-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...