Vinylene carbonate ≥97% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Dioxol-2-one

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

41400-02 41400-04 41400-09
CAAA41400-02EA 53.09 CAD
CAAA41400-02 CAAA41400-04 CAAA41400-09
Vinylene carbonate ≥97% stabilised
Vinylene carbonate

Stab. with BHT


Purity: 97+% CAS No.: 872-36-6 Molecular Formula: C3H2O3 Formula Weight: 86.05 Storage Temperature: 2°C to 8°C Flash Point: 72°(163°F) Physical Form: Liquid Density: 1.355 Refractive Index: 1.4210 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Melting Point: 19-22° Boiling Point: 162-163° MDL No.: MFCD00005380

Formula: C₃H₂O₃
MW: 86.05 g/mol
Boiling Pt: 162…163 °C
Melting Pt: 19…22 °C
Density: 1.355
Flash Pt: 72 °C (163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005380
CAS Number: 872-36-6
EINECS: 212-825-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR