Potassium-tert-pentyloxide 14 - 18% (w/v) in cyclohexane
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium tert-amylate, Potassium 2-methyl-2-butoxide, Potassium tert-pentoxide
41394-14
41394-22
CAAA41394-14EA
176.96
CAD
CAAA41394-14
CAAA41394-22
Potassium-tert-pentyloxide 14 - 18% (w/v) in cyclohexane
Potassium-tert-pentyloxide
Applications: Coreactant for conversion of ketones to oxides
CAS No.: 41233-93-6
Molecular Formula: C5H11KO
Formula Weight: 126.24
Flash Point: -18°(0°F)
Physical Form: Liquid
Density: 0.80
Sensitivity: Air and Moisture Sensitive
UN No.: 2924
MDL No.: MFCD00064808
Applications: Coreactant for conversion of ketones to oxides
Formula:
C₅H₁₁KO MW: 126.24 g/mol Density: 0.80 Flash Pt: –18 °C (0 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064808 CAS Number: 41233-93-6 EINECS: 255-272-5 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...