Chlorhexidine gluconate 20% (w/v) in aqueous solution, not sterile
Supplier: Thermo Scientific Chemicals
41385-AC
41385-AE
41385-AP
41385-AY
CAAA41385-ACEA
60.99
CAD
CAAA41385-AC
CAAA41385-AE
CAAA41385-AP
CAAA41385-AY
Chlorhexidine gluconate 20% (w/v) in aqueous solution, not sterile
Chlorhexidine gluconate
Antiseptic and disinfectant agent
CAS No.: 18472-51-0
Molecular Formula: C22H30Cl2N10.2C6H12O7
Formula Weight: 897.76
Physical Form: Liquid
Density: 1.06
Sensitivity: Light Sensitive
Appearance: Colorless
MDL No.: MFCD00083599
Formula:
C₂₂H₃₀Cl₂N₁₀·2C₆H₁₂O₇ Density: 1.06 Storage Temperature: Ambient |
MDL Number:
MFCD00083599 CAS Number: 18472-51-0 EINECS: 242-354-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...