Octadecylamine hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
41378-06
41378-14
CAAA41378-06EA
73.63
CAD
CAAA41378-06
CAAA41378-14
Octadecylamine hydrochloride
Octadecylamine hydrochloride
CAS No.: 1838-08-0
Molecular Formula: CH3(CH2)17NH2.HCl
Formula Weight: 305.97
Physical Form: Powder
Melting Point: 160°
MDL No.: MFCD00042018
Formula:
C₁₈H₃₉N Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042018 CAS Number: 1838-08-0 EINECS: 217-410-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...