Tungsten zirconium oxide ≥99.7% (metals basis excluding Hf) (max. 0.3% Hf)

Supplier: Thermo Scientific Chemicals

41375-18 41375-30
CAAA41375-18EA 500.86 CAD
CAAA41375-18 CAAA41375-30
Tungsten zirconium oxide ≥99.7% (metals basis excluding Hf) (max. 0.3% Hf)
Tungsten zirconium oxide

Purity: 99.7% CAS No.: 16853-74-0 Molecular Formula: ZrW2O8 Formula Weight: 586.92 Physical Form: -200 Mesh Powder Density: 5.10 Appearance: White to pale green Odor: Odorless MDL No.: MFCD00168096

Formula: ZrW₂O₈
Density: 5.10
Storage Temperature: Ambient
MDL Number: MFCD00168096
CAS Number: 16853-74-0
EINECS: 240-876-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR