Bis(2,2,6,6-tetramethyl-2,5-heptanedionato-O,O')cobalt(II) ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41359-02
41359-04
41359-09
CAAA41359-02EA
104.28
CAD
CAAA41359-02
CAAA41359-04
CAAA41359-09
Bis(2,2,6,6-tetramethyl-2,5-heptanedionato-O,O')cobalt(II) ≥99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-2,5-heptanedionato-O,O')cobalt(II)
Precursor for CVD process
Decomposes at 250°C
Purity: 99.9%
CAS No.: 13986-53-3
Molecular Formula: C22H38CoO4
Formula Weight: 425.46
Physical Form: Powder
Melting Point: 143°
MDL No.: MFCD00233616
Precursor for CVD process
Formula:
C₂₂H₃₈CoO₄ Melting Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233616 CAS Number: 13986-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...