Barium isopropoxide 20% (w/v) in 2-propanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
41338-AB
41338-AD
CAAA41338-ABEA
103.81
CAD
CAAA41338-AB
CAAA41338-AD
Barium isopropoxide 20% (w/v) in 2-propanol
Barium isopropoxide
Applications: In Sol-Gel synthesis and superconductors
CAS No.: 24363-37-9
Molecular Formula: C6H14BaO2
Formula Weight: 255.52
Flash Point: 22°(72°F)
Physical Form: Liquid
Density: 0.89
Appearance: Yellow
Odor: Alcohol-like
UN No.: 3274
MDL No.: MFCD00050486
In Sol-Gel synthesis and superconductors
Formula:
C₆H₁₄BaO₂ Density: 0.89 Flash Pt: 22 °C (72 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00050486 CAS Number: 24363-37-9 UN: 3274 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...