Zinc neodecanoate (17,9 - 18,2% Zn) ≥97%
Supplier: Thermo Scientific Chemicals
41312-30
41312-A1
41312-A7
CAAA41312-30EA
142.04
CAD
CAAA41312-30
CAAA41312-A1
CAAA41312-A7
Zinc neodecanoate (17,9 - 18,2% Zn) ≥97%
Zinc neodecanoate
CAS No.: 27253-29-8
Molecular Formula: Zn(C10H19O2)2
Formula Weight: 407.89
Flash Point: 102°(216°F)
Physical Form: Liquid
Appearance: Pale brown
MDL No.: MFCD00014384
Formula:
C₂₀H₃₈O₄Zn Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014384 CAS Number: 27253-29-8 EINECS: 248-370-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...