Sodium hydroxide monohydrate ≥99.996% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41281-06
41281-14
41281-22
CAAA41281-06EA
54.04
CAD
CAAA41281-06
CAAA41281-14
CAAA41281-22
Sodium hydroxide monohydrate ≥99.996% (metals basis)
Sodium hydroxide monohydrate
Purity: 99.996%
CAS No.: 12179-02-1
Molecular Formula: NaOH.H2O
Formula Weight: 58.01 (40.00anhy)
Physical Form: Crystalline
Density: 2.13
Melting Point: 318.4°
Boiling Point: 1390°
Merck Reference: 14,8627
UN No.: 1823
MDL No.: MFCD00003548
Formula:
HNaO·1H₂O Boiling Pt: 1390 °C Melting Pt: 318.4 °C Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 12179-02-1 EINECS: 215-185-5 UN: 1823 ADR: 8,II Merck Index: 14,8627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...