Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
41279-03
41279-06
41279-14
CAAA41279-03EA
81.53
CAD
CAAA41279-03
CAAA41279-06
CAAA41279-14
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II)
Purity: 99.9%
CAS No.: 14040-05-2
Molecular Formula: C22H38CuO4
Formula Weight: 430.05
Physical Form: Powder
Melting Point: 198°
Boiling Point: 100°/0.1mm subl.
MDL No.: MFCD00058920
Formula:
C₂₂H₃₈CuO₄ MW: 430.08 g/mol Boiling Pt: 100 °C (0.1 mmHg) (subl.) Melting Pt: 198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058920 CAS Number: 14040-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...