Tin(IV) isopropoxide isopropanol adduct ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41269-03
41269-06
CAAA41269-03EA
119.92
CAD
CAAA41269-03
CAAA41269-06
Tin(IV) isopropoxide isopropanol adduct ≥98% (metals basis)
Tin(IV) isopropoxide isopropanol adduct
Purity: 98%
CAS No.: 132951-93-0
Molecular Formula: Sn[OCH(CH3)2]4.C3H7OH
Formula Weight: 414.15
Physical Form: Wet solid
Sensitivity: Moisture Sensitive
Boiling Point: 131°/1.6mm
MDL No.: MFCD00145408
Formula:
Sn[OCH(CH₃)₂]₄·C₃H₇OH MW: 414.15 g/mol Boiling Pt: 131 °C (1.6 mmHg) Melting Pt: ∼ 130 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00145408 CAS Number: 132951-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...