Polyvinyl alcohol, M.W. ∼ 88,000-97,000 87-89%, high molecular weight, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

41240-14 41240-22 41240-36
CAAA41240-14EA 44.56 CAD
CAAA41240-14 CAAA41240-22 CAAA41240-36
Polyvinyl alcohol, M.W. ∼ 88,000-97,000 87-89%, high molecular weight, hydrolysed
Polyvinyl alcohol

Average M.W. 88,000-97,000


CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Physical Form: Solid Odor: Odorless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR