Polyvinyl alcohol, M.W. ∼ 57,000-66,000 86-89%, medium molecular weight, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

41239-14 41239-22 41239-36
CAAA41239-14EA 59.57 CAD
CAAA41239-14 CAAA41239-22 CAAA41239-36
Polyvinyl alcohol, M.W. ∼ 57,000-66,000 86-89%, medium molecular weight, hydrolysed
Polyvinyl alcohol

Average M.W. 57,000-66,000


CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Physical Form: Powder Odor: Odorless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR