Polyvinyl alcohol, M.W. ∼ 10,000-26,000 86-89%, low molecular weight, hydrolysed

Supplier: Thermo Scientific Chemicals

41238-14 41238-22 41238-36
CAAA41238-14EA 58.78 CAD
CAAA41238-14 CAAA41238-22 CAAA41238-36
Polyvinyl alcohol, M.W. ∼ 10,000-26,000 86-89%, low molecular weight, hydrolysed
Polyvinyl alcohol

Average MW: 10,000 to 26,000


CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odor: Odorless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR