Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

41237-AC 41237-AE
CAAA41237-ACEA 195.92 CAD
CAAA41237-AC CAAA41237-AE
Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Cobalt(II)-2-methoxyethoxide

Purity: 99% CAS No.: 142600-61-1 Molecular Formula: C6H14CoO4 Formula Weight: 209.11 Flash Point: 40°(104°F) Physical Form: Liquid Density: 0.96 Sensitivity: Moisture Sensitive Appearance: Dark brown Boiling Point: 124° UN No.: 1188 MDL No.: MFCD00798554

Formula: C₆H₁₄CoO₄
Boiling Pt: 124 °C
Density: 0.96
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00798554
CAS Number: 142600-61-1
UN: 1188
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR