[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex (1:1) (13% Pd)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex

41225-03 41225-06 41225-14
CAAA41225-03EA 104.28 CAD
CAAA41225-03 CAAA41225-06 CAAA41225-14
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex (1:1) (13% Pd)
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride dichloromethane complex

CAS No.: 95464-05-4 Molecular Formula: C34H28Cl2FeP2Pd.CH2Cl2 Formula Weight: 816.64 Physical Form: Powder Appearance: Orange-red Odor: Nearly odorless MDL No.: MFCD00792899

Formula: C₃₅H₃₀Cl₄FeP₂Pd
MW: 816.64 g/mol
Melting Pt: 275…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00792899
CAS Number: 95464-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR