α-Naphtholphthalein
Supplier: Thermo Scientific Chemicals
|
Warning
|
41221-03
41221-06
41221-14
CAAA41221-03EA
79.63
CAD
CAAA41221-03
CAAA41221-06
CAAA41221-14
α-Naphtholphthalein
α-Naphtholphthalein
Applications: Indicator in 0.1% or 0.04% alcohol solution
pH 7.3 (colorless to red), pH 8.7 (green to blue)
CAS No.: 596-01-0
Molecular Formula: C28H18O4
Formula Weight: 418.45
Physical Form: Powder
Appearance: Pale brown
Melting Point: 238-240°
MDL No.: MFCD00036202
Indicator in 0.1% or 0.04% alcohol solution
Formula:
C₂₈H₁₈O₄ Melting Pt: 238…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036202 CAS Number: 596-01-0 EINECS: 209-875-5 Merck Index: 14,6389 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...