Di-sec-butoxyaluminoxytriethoxysilane

Supplier: Thermo Scientific Chemicals
Warning

41210-09 41210-18 41210-30
CAAA41210-09EA 74.26 CAD
CAAA41210-09 CAAA41210-18 CAAA41210-30
Di-sec-butoxyaluminoxytriethoxysilane
Di-sec-butoxyaluminoxytriethoxysilane

CAS No.: 68959-06-8 Molecular Formula: C14H33AlO6Si Formula Weight: 352.48 Flash Point: 46°(115°F) Physical Form: Liquid Density: 1.00 Sensitivity: Moisture Sensitive Appearance: Colorless Boiling Point: 200° UN No.: 1993 MDL No.: MFCD00053771

Formula: C₁₄H₃₃AlO₆Si
Boiling Pt: >200 °C
Density: 1,00
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053771
CAS Number: 68959-06-8
EINECS: 273-392-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR