Gold Palladium (60:40) (w/w) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
41206-BU
41206-BW
41206-G1
CAAA41206-BUEA
161.16
CAD
CAAA41206-BU
CAAA41206-BW
CAAA41206-G1
Gold Palladium (60:40) (w/w) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Gold Palladium
Purity: 99.9%
Molecular Formula: Au:Pd; 60:40 wt%
Physical Form: Temper: soft est .154g/ft
MDL No.: MFCD00003436
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...