Fullerene, hydroxylated, C60(OH)n, powder

Supplier: Thermo Scientific Chemicals
Warning

41182-MB 41182-MC
CAAA41182-MBEA 300.2 CAD
CAAA41182-MB CAAA41182-MC
Fullerene, hydroxylated, C60(OH)n, powder
Fullerene, hydroxylated, C60(OH)n

UN No.: 1325 MDL No.: MFCD06202029

Storage Temperature: Ambient
MDL Number: MFCD06202029
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR