Tellurium(IV) ethoxide ≥85% remainder ethanol, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
41105-04
41105-09
CAAA41105-04EA
197.5
CAD
CAAA41105-04
CAAA41105-09
Tellurium(IV) ethoxide ≥85% remainder ethanol, tech.
Tellurium(IV) ethoxide
Contains 10% ethanol.
Decomposes in water to TeO2
Purity: 85%
CAS No.: 2017-01-8
Molecular Formula: Te(OC2H5)4
Formula Weight: 307.84
Flash Point: 27°(81°F)
Physical Form: Liquid
Density: 1.52
Sensitivity: Moisture and Light Sensitive
Odor: Alcohol-like
Melting Point:
Formula:
Te(OC₂H₅)₄ Boiling Pt: 88…90 °C (2 mmHg) Melting Pt: >100 °C (dec.) Density: 1,52 Flash Pt: 27 °C (81 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00270937 CAS Number: 2017-01-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...