Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxideSilicium (IV) oxideSilicium dioxide

41099-KS 41099-KS
CAAA41099-KSEA 714.16 CAD
AA41099-KS CAAA41099-KS
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Silicium dioxide
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1719 °C
Density: 2.2 g/cm³ (20 °C)
MDL Number: MFCD00011232
CAS Number: 7631-86-9
Merck Index: 14,08493

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR