Zirconium disulfate tetrahydrate 98+% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

41041-22 41041-36 41041-A3
CAAA41041-22EA 73.31 CAD
CAAA41041-22 CAAA41041-36 CAAA41041-A3
Zirconium disulfate tetrahydrate 98+% (metals basis)
Zirconium disulphate tetrahydrate

Catalyst support
Soluble in water


Purity: 98+% CAS No.: 7446-31-3 Molecular Formula: Zr(SO4)2.4H2O Formula Weight: 355.40 (283.34anhy) Physical Form: -20 Mesh Density: 3.22 Appearance: White Solubility: Soluble in water Merck Reference: 14,10182 MDL No.: MFCD00149267


Catalyst support

Formula: O₈S₂Zr·4H₂O
Density: 3.22
Storage Temperature: Ambient
MDL Number: MFCD00149267
CAS Number: 7446-31-3
EINECS: 238-694-4
Merck Index: 14,10182

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR