Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

41039-G1 41039-G5 41039-G6
CAAA41039-G1EA 116.92 CAD
CAAA41039-G1 CAAA41039-G5 CAAA41039-G6
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.05 mm (0.002 in)
Tungsten (gold coated)

Purity: 99.95% Physical Form: ≈0.02g/m (W only), 3-5 wt% Au plating MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR