Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

41038-G1 41038-G5 41038-G6
CAAA41038-G1EA 141.41 CAD
CAAA41038-G1 CAAA41038-G5 CAAA41038-G6
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.025 mm (0.001 in)
Tungsten (gold coated)

Uses: Microelectronics, evaporation source, high temperature


Purity: 99.95% Physical Form: ≈0.005g/m (W only), 3-5 wt% Au plating Odor: Odorless MDL No.: MFCD00011461


Microelectronics, evaporation source, high temperature

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR