Lead tin, solder alloy, with rosin core Pb:Sn - 50:50 %, Ø 1.6 mm (0.06 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
41024-29
41024-TB
CAAA41024-29EA
159.58
CAD
CAAA41024-29
CAAA41024-TB
Lead tin, solder alloy, with rosin core Pb:Sn - 50:50 %, Ø 1.6 mm (0.06 in)
Lead tin, solder alloy, with rosin core
Melting temperature range: 182-215°
Molecular Formula: Pb:Sn; 50:50 wt%
MDL No.: MFCD00798549
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...