Amberlite® IRN-150, Amberlite™ ion exchange resin
Supplier: Thermo Scientific Chemicals
Synonyms:
Styrene-DVB
40992-36
40992-A3
40992-A9
CAAA40992-36EA
79.79
CAD
CAAA40992-36
CAAA40992-A3
CAAA40992-A9
Amberlite® IRN-150, Amberlite™ ion exchange resin
Amberlite® IRN-150
Uses: Water treatment and purification
Molecular Formula: Styrene-DVB
Odor: Odorless
MDL No.: MFCD00145822
Applications: Water treatment and purification
Storage Temperature:
Ambient |
MDL Number:
MFCD00145822 CAS Number: 79956-14-2 Merck Index: 14,382 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...