Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.51 mm (0.02 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
40946-BW
40946-BZ
CAAA40946-BWEA
54.83
CAD
CAAA40946-BW
CAAA40946-BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.51 mm (0.02 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odor: Odorless
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...