Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.25 mm (0.01 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
40945-BW
40945-BZ
CAAA40945-BWEA
44.71
CAD
CAAA40945-BW
CAAA40945-BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.25 mm (0.01 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odor: Odorless
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...