Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.127 mm (0.005 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
40944-BW
40944-BZ
CAAA40944-BWEA
44.24
CAD
CAAA40944-BW
CAAA40944-BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.127 mm (0.005 in) Type 304
Stainless steel
Uses: Filtration, electrodes
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odor: Odorless
MDL No.: MFCD00148499
Applications: Filtration, electrodes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...