Hastelloy C® alloy, gauze 100 mesh woven from wire 0.102 mm (0.004in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40940-FL
40940-GT
40940-HB
CAAA40940-FLEA
83.27
CAD
CAAA40940-FL
CAAA40940-GT
CAAA40940-HB
Hastelloy C® alloy, gauze 100 mesh woven from wire 0.102 mm (0.004in)
Hastelloy C® alloy
Physical Form: Wire Cloth
MDL No.: MFCD00801065
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...