Hastelloy C® alloy, gauze woven from wire, Hastelloy C® 20 mesh woven from 0.23mm (0.009in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40938-FL
40938-GT
40938-HB
CAAA40938-FLEA
64.15
CAD
CAAA40938-FL
CAAA40938-GT
CAAA40938-HB
Hastelloy C® alloy, gauze woven from wire, Hastelloy C® 20 mesh woven from 0.23mm (0.009in)
Hastelloy C® alloy
Applications: High strength, corrosion resistant alloy
Physical Form: Wire Cloth
MDL No.: MFCD00801065
Applications: High strength, corrosion resistant alloy
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...