Tungsten (gold coated), gauze woven from wire 50 mesh, Ø 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

40926-FL 40926-GT 40926-HB
CAAA40926-FLEA 565.64 CAD
CAAA40926-FL CAAA40926-GT CAAA40926-HB
Tungsten (gold coated), gauze woven from wire 50 mesh, Ø 0.025 mm (0.001 in)
Tungsten (gold coated)

Uses: Filtration, electrodes
Open area: 90.3%; Width of opening: 0.019in


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461


Filtration, electrodes

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR