Nickel(II) oxalate hydrate ≥99.9985%, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
40757-06
40757-14
CAAA40757-06EA
60.36
CAD
CAAA40757-06
CAAA40757-14
Nickel(II) oxalate hydrate ≥99.9985%, Puratronic®
Nickel(II) oxalate hydrate
Purity: 99.9985%
CAS No.: 126956-48-7
Molecular Formula: C2NiO4.xH2O (x≈2)
Formula Weight: 146.73(anhy)
Physical Form: Crystalline powder
Appearance: Green
UN No.: 3077
MDL No.: MFCD00167155
Formula:
C₂NiO₄·xH₂O(x≈2) MW: 146.73 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00167155 CAS Number: 126956-48-7 EINECS: 208-933-7 UN: 3077 ADR: 9 III Merck Index: 14,6514 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...