Zinc tert-butoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
40718-03
40718-MD
CAAA40718-03EA
470.84
CAD
CAAA40718-03
CAAA40718-MD
Zinc tert-butoxide
Zinc tert-butoxide
CAS No.: 4278-43-7
Molecular Formula: C8H18O2Zn
Formula Weight: 211.61
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odor: Odorless
UN No.: 3181
MDL No.: MFCD00145545
Formula:
C₈H₁₈O₂Zn Storage Temperature: Ambient |
MDL Number:
MFCD00145545 CAS Number: 4278-43-7 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...