Molybdenum, gold plated ≥99.95% (metals basis), wire, Ø 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
40671-G5
40671-G6
CAAA40671-G5EA
379.2
CAD
CAAA40671-G5
CAAA40671-G6
Molybdenum, gold plated ≥99.95% (metals basis), wire, Ø 0.025 mm (0.001 in)
Molybdenum, gold plated
Uses: Microelectronics, high temperature
Purity: 99.95%
CAS No.: 7439-98-7
Physical Form: ≈0.005g/m (Mo only), 3-5 wt% Au plating
MDL No.: MFCD00003465
Applications: Microelectronics, high temperature
Storage Temperature:
Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...