Samarium(III) 2,4-pentanedionate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
40665-06
40665-14
CAAA40665-06EA
59.25
CAD
CAAA40665-06
CAAA40665-14
Samarium(III) 2,4-pentanedionate hydrate ≥99.9%
Samarium(III) 2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 64438-53-5
Molecular Formula: C15H21O6Sm.xH2O
Formula Weight: 447.68(anhy)
Physical Form: Powder
MDL No.: MFCD00792904
Formula:
C₁₅H₂₁O₆Sm.xH₂O MW: 447.68 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00792904 CAS Number: 64438-53-5 EINECS: 238-632-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...