Yttrium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

40651-18 40651-30
CAAA40651-18EA 135.88 CAD
CAAA40651-18 CAAA40651-30
Yttrium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) carbonate hydrate

Purity: 99.9% CAS No.: 38245-39-5 Molecular Formula: Y2(CO3)3.xH2O Formula Weight: 357.84(anhy) Physical Form: Powder MDL No.: MFCD00149943

Formula: Y₂(CO₃)₃·xH₂O
MW: 357.84 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149943
CAS Number: 38245-39-5
EINECS: 209-119-4
Merck Index: 14,10107

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR