Neodymium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perchloric acid neodymium salt hexahydrate

40568-NB 40568-ND
CAAA40568-NBEA 108.55 CAD
CAAA40568-NB CAAA40568-ND
Neodymium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Neodymium(III) perchlorate hexahydrate

CAS No.: 17522-69-9 Molecular Formula: Nd(ClO4)3.6H2O Formula Weight: 550.68 (442.59anhy) Physical Form: Liquid Odor: Odorless UN No.: 3211 MDL No.: MFCD00198195

Formula: Nd(ClO4)3.6H2O
MW: 550.68 g/mol (442.59 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00198195
CAS Number: 17522-69-9
EINECS: 236-821-8
UN: 3211
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR