Polyethyleneimine, M.W. 600 ≥99%, branched

Supplier: Thermo Scientific Chemicals
Warning

40527-14 40527-22 40527-36
CAAA40527-14EA 80.74 CAD
CAAA40527-14 CAAA40527-22 CAAA40527-36
Polyethyleneimine, M.W. 600 ≥99%, branched
Polyethyleneimine

Purity: 99% CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Pale yellow Odor: Amine-like MDL No.: MFCD00803910

Formula: (CH2CH2NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR