Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40524-02
40524-04
CAAA40524-02EA
213.3
CAD
CAAA40524-02
CAAA40524-04
CAAA40524-09
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Purity: 99%
CAS No.: 25360-32-1
Molecular Formula: C55H47OP3Ru
Formula Weight: 917.97
Physical Form: Crystalline
Sensitivity: Moisture and Light Sensitive
Appearance: White
Melting Point: 161-163°
MDL No.: MFCD00015870
Formula:
C₅₅H₄₇OP₃Ru MW: 917.97 g/mol Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015870 CAS Number: 25360-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...