Chromyl chloride ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Applications: Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
Purity: 99.99%
CAS No.: 14977-61-8
Molecular Formula: CrO2Cl2
Formula Weight: 154.90
Physical Form: Liquid
Density: 1.911
Sensitivity: Moisture Sensitive
Odor: Pungent, makes eyes water
Melting Point: -96.5°
Boiling Point: 117°
UN No.: 1758
MDL No.: MFCD00010951
Applications: Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
Formula:
CrO₂Cl₂ Boiling Pt: 117 °C Melting Pt: –96.5 °C Density: 1.911 Storage Temperature: Ambient |
MDL Number:
MFCD00010951 CAS Number: 14977-61-8 EINECS: 239-056-8 UN: 1758 ADR: 8,I Merck Index: 14,2248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...