Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), Ni(TMHD)2
40470-03
40470-06
CAAA40470-03EA
181.7
CAD
CAAA40470-03
CAAA40470-06
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II)
Applications: NMR shift reagent, nickel oxide precursor
Purity: 98%
CAS No.: 14481-08-4
Molecular Formula: C22H38NiO4
Formula Weight: 425.24
Physical Form: Powder
Melting Point: 223-225°
Boiling Point: 90-110°/0.1mm subl.
MDL No.: MFCD00192348
NMR shift reagent, nickel oxide precursor
Formula:
C₂₂H₃₈NiO₄ MW: 425.23 g/mol Boiling Pt: 90…110 °C (0.1 mmHg) (subl.) Melting Pt: 223…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192348 CAS Number: 14481-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...